Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Bromofluorene, 97+%

Catalog No. p-7080756
Click to view available options
Quantity:
5 g
50 g

CAS: 1940-57-4 | C13H9Br | 245.119 g/mol

9-Bromofluorene, is used to treat skin dermatological conditions and in pharmaceutical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9-Bromofluorene, is used to treat skin dermatological conditions and in pharmaceutical industry.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents. Air sensitive. Stable under normal conditions. Preserve in light-resistant and well-closed containers
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1940-57-4
Molecular Formula C13H9Br
Molecular Weight (g/mol) 245.119
MDL Number MFCD00001133
InChI Key AHCDKANCCBEQJJ-UHFFFAOYSA-N
Synonym 9-bromofluorene, fluorene, 9-bromo, 9h-fluorene, 9-bromo, 9-fluorenyl bromide, monobromofluorene, 9-bromo-fluoren, 9-brom-9h-fluoren, acmc-1bqkc, 9-bromanyl-9h-fluorene
PubChem CID 16024
IUPAC Name 9-bromo-9H-fluorene
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)Br

Specifications

Melting Point 101°C to 105°C
Odor Irritating
Quantity 5 g
UN Number UN3261
Beilstein 2047220
Solubility Information Insoluble in water.
Formula Weight 245.12
Percent Purity ≥98%
Chemical Name or Material 9-Bromofluorene
Hazard Category H314-H317
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P272-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 217-722-9
RTECSNumber LL5890000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.