Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Chloroanthracene, 96%, Thermo Scientific Chemicals

Catalog No. p-7043514
Click to view available options
Quantity:
5 g

9-Chloroanthracene, is used as a chlorinated anthracene compound used for biochemical research. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9-Chloroanthracene, is used as a chlorinated anthracene compound used for biochemical research. It is also used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 716-53-0
Molecular Formula C14H9Cl
Molecular Weight (g/mol) 212.676
MDL Number MFCD00001245
InChI Key KULLJOPUZUWTMF-UHFFFAOYSA-N
PubChem CID 69744
IUPAC Name 9-chloroanthracene
SMILES C1=CC=C2C(=C1)C=C3C=CC=CC3=C2Cl

Specifications

Melting Point 101°C to 106°C
Quantity 5 g
Beilstein 1869540
Synonym anthracene, 9-chloro, ccris 5547, 9-chloro-anthracene, #, anthracene,9-chloro, bidd:gt0730
Solubility Information Insoluble in water.
Formula Weight 212.68
Percent Purity 96%
Chemical Name or Material 9-Chloroanthracene
Hazard Category H315-H319-H335-H341
Hazard Statement GHS H Statement
H341-H315-H319-H335
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c
EINECSNumber 211-937-1
RTECSNumber CA9561000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.