Learn More
9-Decen-1-ol, 90+%
Description
It finds it uses in a wide range of fragrances, especially in fine fragrances and soaps, in floral, rose petal or herbal fragrances. 9-Decen-1-ol is used in the preparation of semifluorinated acids required for the synthesis of poly(styrene-b-semi fluorinated isoprene) block copolymers with -CF2H-terminated side groups.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It finds it uses in a wide range of fragrances, especially in fine fragrances and soaps, in floral, rose petal or herbal fragrances. 9-Decen-1-ol is used in the preparation of semifluorinated acids required for the synthesis of poly(styrene-b-semi fluorinated isoprene) block copolymers with -CF2H-terminated side groups.
Solubility
Soluble in water at 20°C 0.16g/L. Soluble in alcohol, dipropylene glycol, paraffin oil.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Specifications
| Melting Point | -13°C |
| Density | 0.847 |
| Boiling Point | 235°C |
| Flash Point | 104°C (219°F) |
| Odor | Pleasant |
| Refractive Index | 1.448 |
| Quantity | 25 g |
| Beilstein | 1750928 |
| Solubility Information | Soluble in water at 20°C 0.16g/L. Soluble in alcohol,dipropylene glycol,paraffin oil. |
| Formula Weight | 156.27 |
| Show More |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319 Causes skin irritation. Causes serious eye irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 235-878-6 |
| RTECSNumber | HE2095000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.