Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Decenoic Acid 98.0+%, TCI America™
SDP

Catalog No. d19325ml Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 mL
25 mL

Chemical Identifiers

CAS 14436-32-9
Molecular Formula C10H18O2
Molecular Weight (g/mol) 170.252
MDL Number MFCD00036663
InChI Key KHAVLLBUVKBTBG-UHFFFAOYSA-N
PubChem CID 61743
ChEBI CHEBI:32381
IUPAC Name dec-9-enoic acid
SMILES C=CCCCCCCCC(=O)O

Specifications

Color Yellow
Boiling Point 162°C
Quantity 5 mL
Formula Weight 170.25
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 9-Decenoic Acid
EINECSNumber (2)-0609
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.