Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Decenyl Acetate 97.0+%, TCI America™
SDP

Catalog No. A109325ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A109325ML 25 mL
A10935ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A109325ML Supplier TCI America Supplier No. A109325ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 50816-18-7
Molecular Formula C12H22O2
Molecular Weight (g/mol) 198.306
MDL Number MFCD00036512
InChI Key PIQXMYAEJSMANF-UHFFFAOYSA-N
Synonym Acetic Acid 9-Decenyl Ester, 10-Acetoxy-1-decene
PubChem CID 39801
IUPAC Name dec-9-enyl acetate
SMILES CC(=O)OCCCCCCCCC=C

Specifications

Color Colorless
Boiling Point 246°C
Quantity 25 mL
Formula Weight 198.31
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 9-Decenyl Acetate
EINECSNumber (2)-0762
RTECSNumber AG5230000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.