Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Fluorenemethanol, 99%

Catalog No. p-3757932
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
50 g
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC170930010 1 g Glass bottle
AC170930050 5 g Glass bottle
AC170930100 10 g Glass bottle
AC170930500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC170930010 Supplier Thermo Scientific Chemicals Supplier No. 170930010
Only null left
Add to Cart

CAS: 24324-17-2 | C14H12O | 196.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24324-17-2
Molecular Formula C14H12O
Molecular Weight (g/mol) 196.25
MDL Number MFCD00001139
InChI Key XXSCONYSQQLHTH-UHFFFAOYSA-N
Synonym 9-fluorenemethanol, 9-fluorenylmethanol, 9h-fluorene-9-methanol, 9-fluorenyl methanol, 9h-fluoren-9-yl methanol, fluorene-9-methanol, 9-hydroxymethylfluorene, 9-fluorenmethanol, hofm, fluoren-9-yl methanol
PubChem CID 90466
SMILES OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 104°C to 107°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 1 g
Beilstein 06, III, 3507
Fieser 05,308; 03,145; 04,237; 13,134
IUPAC Name (9H-fluoren-9-yl)methanol
Formula Weight 196.25
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 9-Fluorenemethanol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
EINECSNumber 246-167-5

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.