Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Methylanthracene, 99%

Catalog No. p-7022786
Click to view available options
Quantity:
1 g
5 g
25 g

CAS: 779-02-2 | C15H12 | 192.26 g/mol

9-Methylanthracene used as a Intermediates for pharmaceutical and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9-Methylanthracene used as a Intermediates for pharmaceutical and chemical research.

Solubility
Soluble in water (0,0003 mg/L at 25°C) and toluene. Slightly soluble in benzene.

Notes
Mechanical exhaust required. When not in use, tightly seal the container and store in a dry, cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 779-02-2
Molecular Formula C15H12
Molecular Weight (g/mol) 192.26
MDL Number MFCD00001261
InChI Key CPGPAVAKSZHMBP-UHFFFAOYSA-N
Synonym anthracene, 9-methyl, unii-65nk4cin03, ccris 2740, 9-methyl anthracene, 9-methyl-anthracene, anthracen-9-ylmethyl, acmc-209pcm, 9-methylanthracene, wln: l c666j b1, 9-methylanthracene, analytical standard
PubChem CID 13068
IUPAC Name 9-methylanthracene
SMILES CC1=C2C=CC=CC2=CC2=CC=CC=C12

Specifications

Melting Point 77°C to 79°C
Density 1.066
Boiling Point 196°C to 197°C (16 mmHg)
Quantity 1 g
UN Number UN3077
Beilstein 1907463
Solubility Information Soluble in water (0,0003mg/L at 25°C) and toluene. Slightly soluble in benzene.
Formula Weight 192.26
Percent Purity 99%
Chemical Name or Material 9-Methylanthracene
Hazard Category H318
Hazard Statement GHS H Statement
H318
Causes serious eye damage.
Precautionary Statement P280-P305+P351+P338-P310
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 212-299-7
RTECSNumber CB0700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.