Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Phenanthrenecarboxaldehyde, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
4707-71-5
C15H10O
206.244
MFCD00001175
QECIGCMPORCORE-UHFFFAOYSA-N
9-phenanthrenecarboxaldehyde,phenanthrene-9-carboxaldehyde,9-formylphenanthrene,phenanthrene-9-aldehyde,9-phenanthrenecarbaldehyde,9-phenanthrenealdehyde,9-phenanthrencarbaldehyd,9-phenanthraldehyde,9-phenanthrenaldehyde,phenanthren-9-aldehyd
78437
phenanthrene-9-carbaldehyde
C1=CC=C2C(=C1)C=C(C3=CC=CC=C23)C=O
4707-71-5
C15H10O
206.244
MFCD00001175
QECIGCMPORCORE-UHFFFAOYSA-N
9-phenanthrenecarboxaldehyde,phenanthrene-9-carboxaldehyde,9-formylphenanthrene,phenanthrene-9-aldehyde,9-phenanthrenecarbaldehyde,9-phenanthrenealdehyde,9-phenanthrencarbaldehyd,9-phenanthraldehyde,9-phenanthrenaldehyde,phenanthren-9-aldehyd
78437
phenanthrene-9-carbaldehyde
C1=CC=C2C(=C1)C=C(C3=CC=CC=C23)C=O
Melting Point 99°C to 103°C
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 1g
Color Yellow
Formula Weight 206.24
Physical Form Powder
Percent Purity ≥96.0%
Chemical Name or Material 9-Phenanthrenecarboxaldehyde

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.