Learn More
R&D Systems™ 9-Phenanthrol
SDP
Catalog No. 499950R Shop All Novus Products
Change view
Click to view available options
Quantity:
50 mg
Catalog No. Quantity
49-995-0R 50 mg
1 options
This item is not returnable. View return policy
Catalog No. 49-995-0R Supplier R&D Systems™ Supplier No. 4999/50
Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.
This item is not returnable. View return policy
A Tocris product, part of R&D Systems. Selective TRPM4 blocker
9-Phenanthrol is a selective TRPM4 blocker (IC50 = 20 μM in HEK293 cells). Exhibits no effect on CFTR or TRPM5 (at 0.25 and 1 mM respectively). Abolishes arrhythmias induced by hypoxia in a mouse heart model.
Specifications
Quantity 50 mg
Product Type 9-Phenanthrol
Molecular Weight (g/mol) 194.23
Product Line Tocris
Inhibitors 9-Phenanthrol