Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Phenyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole 98.0+%, TCI America™
SDP

Catalog No. p-7135542
Click to view available options
Quantity:
200 mg
1 g

Chemical Identifiers

CAS 1246669-45-3
Molecular Formula C24H24BNO2
Molecular Weight (g/mol) 369.27
MDL Number MFCD28129441
InChI Key WUXIRZBTTICLCG-UHFFFAOYSA-N
Synonym 9-Phenylcarbazole-2-boronic Acid Pinacol Ester, 4,4,5,5-Tetramethyl-2-(9-phenylcarbazol-2-yl)-1,3,2-dioxaborolane
PubChem CID 59858738
IUPAC Name 9-phenyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole
SMILES CC1(C)OB(OC1(C)C)C1=CC2=C(C=C1)C1=C(C=CC=C1)N2C1=CC=CC=C1

Specifications

Melting Point 149°C
Color White
Quantity 200 mg
Formula Weight 369.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9-Phenyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.