Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Phenylanthracene 98.0+%, TCI America™
SDP

Catalog No. P013825G
Click to view available options
Quantity:
1 g
5 g
25 g

Chemical Identifiers

CAS 602-55-1
Molecular Formula C20H14
Molecular Weight (g/mol) 254.33
MDL Number MFCD00001252
InChI Key LUBXLGUQZVKOFP-UHFFFAOYSA-N
Synonym anthracene, 9-phenyl, 9-phenyl-anthracene, 10-phenylanthracene, 9-phenyl-anthracen, 9-phenylanthracene, acmc-1b14f, anthracene, 9-phenyl-8ci 9ci
PubChem CID 11766
IUPAC Name 9-phenylanthracene
SMILES C1=CC=C(C=C1)C1=C2C=CC=CC2=CC2=CC=CC=C12

Specifications

Melting Point 154°C
Quantity 25 g
Formula Weight 254.33
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9-Phenylanthracene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.