Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Vinylanthracene, 97%

Catalog No. p-7026368
Click to view available options
Quantity:
1 g
5 g

CAS: 2444-68-0 | C16H12 | 204.272 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2444-68-0
Molecular Formula C16H12
Molecular Weight (g/mol) 204.272
MDL Number MFCD00003576
InChI Key OGOYZCQQQFAGRI-UHFFFAOYSA-N
Synonym 9-vinylanthracene, anthracene, 9-ethenyl, unii-a3692nge8j, 9-vinyl-anthracene, 9-ethenyl anthracene, 9-ethenylanthracen, 10-vinylanthracene
PubChem CID 17125
IUPAC Name 9-ethenylanthracene
SMILES C=CC1=C2C=CC=CC2=CC3=CC=CC=C31

Specifications

Melting Point 62°C to 65°C
Boiling Point 61°C to 66°C (10 mmHg)
Quantity 1 g
Beilstein 2041641
Formula Weight 204.27
Percent Purity 97%
Chemical Name or Material 9-Vinylanthracene
EINECSNumber 219-486-2
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.