Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GVS Magna™ 90 mm Nylon Disc

Catalog No. 09735238 Shop All Osmonics Products
Change view
Click to view available options
Pore Size:
0.8 μm
1.2 μm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pore Size
09-735-238 1.2 μm
09-924-052 0.8 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 09-735-238 Supplier GVS Magna™ Supplier No. 1213798
Add to Cart
Add to Cart

A supported, naturally hydrophilic membrane designed to wet out evenly and retain its superior strength during use in general filtration or medical assays.

Properties

  • Internally supported with an inert polyester support web giving it added dimensional strength and stability that prevents cracking, tearing, curling and breaking.
  • This added strength and durability is advantageous during usage that involves aggressive handling or automated equipment.
  • A naturally hydrophilic membrane, does not require wetting agents that can interfere with biological processes.
  • This membrane has porosity of 70% to 80% for high diffusion and low-flow resistance.

Characteristics

  • Hydrophilic eliminates the need for wetting agents that can potentially interfere with biological processes.
  • Super strength eases handling when used with automated equipment.
  • Low extractables ensures tests will be clean and pure leading to more consistent results.
  • Lot-to-lot consistency quality checks ensure lot-to-lot consistency, both down and across the polyester web, for dependable results every time.
  • Thickness: 0.65 to 0.125 mm
  • Sterilization: Autoclavable

Applications

  • HPLC sample preparation
  • Sterilization and clarification of aqueous and organic solvent solutions

Specifications

Autoclavable Autoclavable
Disposable Limited-use
Filter Type Membrane Filter
Format Disc
Material Nylon
Pore Size 1.2 μm
Sterility Non-sterile
Type Disc Membrane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.