Learn More
Invitrogen™ 90K/Mac-2BP Monoclonal Antibody (SP-2), eBioscience™
Mouse Monoclonal Antibody
Supplier: Invitrogen™ BMS146

Description
Description: BMS146 specifically recognizes Mac-2 (galectin-3) mucin-like binding domain of human 90K/Mac-2BP. Mac-2BP is a highly glycosylated protein that plays a major role in cancer research. Elevated levels in sera have been found in many patients suffering from various types of cancer or viral infections. Applications Tested: Immunohistochemistry (Paraffin), Western Blotting. Purity: >95%.
The galectins are a family of beta-galactoside-binding proteins implicated in modulating cell-cell and cell-matrix interactions. LGALS3BP has been found elevated in the serum of patients with cancer and in those infected by the human immunodeficiency virus (HIV). It appears to be implicated in immune response associated with natural killer (NK) and lymphokine-activated killer (LAK) cell cytotoxicity. Using fluorescence in situ hybridization the full length 90K cDNA has been localized to chromosome 17q25. The native protein binds specifically to a human macrophage-associated lectin known as Mac-2 and also binds galectin 1.
Specifications
90K/Mac-2BP | |
Monoclonal | |
0.1 mg/mL | |
PBS with no preservative; pH 7.2-7.4 | |
Q08380 | |
LGALS3BP | |
Rec. Human Mac-2BP. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry, Western Blot | |
SP-2 | |
Unconjugated | |
LGALS3BP | |
90K; basement membrane autoantigen p105; BTBD17B; CyCAP; Cyp-C-associated protein; galectin 3 binding protein; galectin-3-binding protein; gp90; L3 antigen; Lectin; lectin galactoside-binding soluble 3 binding protein; lectin galactoside-binding soluble 3-binding protein; lectin, galactoside binding soluble 3 binding protein; lectin, galactoside-binding, soluble, 3 binding protein; Lgals3bp; M2BP; mac-2 BP; mac-2-binding protein; MAC2BP; MAC-2BP; MAC-2-BP; mama; peptidylprolyl isomerase C-associated protein; Ppicap; Protein 90K; Protein MAMA; serum protein 90K; similar to Galectin-3 binding protein precursor (Lectin galactoside-binding soluble 3 binding protein) (Mac-2 binding protein) (Mac-2 BP) (MAC2BP) (Tumor-associated antigen 90K); TANGO10B; transport and golgi organization 10 homolog B; tumor-associated antigen 90K | |
Mouse | |
Affinity Chromatography | |
RUO | |
3959 | |
4°C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.