Learn More
Invitrogen™ 90K/Mac-2BP Monoclonal Antibody (SP-2), FITC, eBioscience™

Description
BMS146 specifically recognizes Mac-2 (galectin-3) mucin-like binding domain of human 90K/Mac-2BP. Mac-2BP is a highly glycosylated protein that plays a major role in cancer research. Elevated levels in sera have been found in many patients suffering from various types of cancer or viral infections. Purity: >95%. Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.
Specifications
Specifications
| Antigen | 90K/Mac-2BP |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | SP-2 |
| Concentration | 10 μL/Test |
| Conjugate | FITC |
| Formulation | TBS with 1% BSA and 0.02% sodium azide; pH 7.2-7.4 |
| Gene | LGALS3BP |
| Gene Accession No. | Q08380 |
| Gene Alias | 90K; basement membrane autoantigen p105; BTBD17B; CyCAP; Cyp-C-associated protein; galectin 3 binding protein; galectin-3-binding protein; gp90; L3 antigen; Lectin; lectin galactoside-binding soluble 3 binding protein; lectin galactoside-binding soluble 3-binding protein; lectin, galactoside binding soluble 3 binding protein; lectin, galactoside-binding, soluble, 3 binding protein; Lgals3bp; M2BP; mac-2 BP; mac-2-binding protein; MAC2BP; MAC-2BP; MAC-2-BP; mama; peptidylprolyl isomerase C-associated protein; Ppicap; Protein 90K; Protein MAMA; serum protein 90K; similar to Galectin-3 binding protein precursor (Lectin galactoside-binding soluble 3 binding protein) (Mac-2 binding protein) (Mac-2 BP) (MAC2BP) (Tumor-associated antigen 90K); TANGO10B; transport and golgi organization 10 homolog B; tumor-associated antigen 90K |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.