Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One 96-well, Advanced TC Treated, Flat-Bottom Microplate
GREENER_CHOICE

Catalog No. p-9045062
Click to view available options
Packaging:
1/Pk., 100/Cs.

Color: Clear; Sterility: Sterile; Lid: With Lid

Advanced TC polymer modification changes plastic surface in a way that increases primary and long-term cell adhesion and positively influences cell-dependant features and function enhanced cell attachment and higher proliferation rates facilitate rapid cell expansion even under restricted growth conditions.
  • Provide conditions for improved cell adherence, accelerated cell expansion and increase overall cell yield
  • For use with serum reduced and serum-free media
  • Allows differentiation of semi-adherent cells
  • Non-pyrogenic and non-cytotoxic
  • Homogenous cell growth
  • Higher transgene activity
TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Lid With Lid
Culture Area 34 mm2
Array 8 x 12
Packaging 1/Pk., 100/Cs.
Color Clear
Material Polystyrene
Volume (Metric) Well 25 to 340 μL
No. of Wells 96
Sterility Sterile
Surface Treatment Advanced TC
Well Shape Flat Bottom (F-well)
No. per Case 100
Quantity 100/Cs.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.