Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ 96-Well, Cell Culture-Treated, Flat-Bottom Microplate
BIOPHARMAGSA_VAGREENER_CHOICE

Catalog No. p-9045029
Click to view available options
Packaging:
20/Pk., 100/Cs.
Individually wrapped, 48/Cs.
This item is not returnable. View return policy
This item is not returnable. View return policy

Color: Black, Clear Bottom; Sterility: Sterile; Lid: With Lid

  • Bottoms are 60% thinner than conventional polystyrene microplates, resulting in lower background fluorescence and enabling readings down to 340 nm.
  • Opaque walls prevent well-to-well cross-talk.
  • Optically clear flat bottom permits direct microscopic viewing.
  • This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

    TRUSTED_SUSTAINABILITY

    Specifications

    Format Standard
    Lid With Lid
    Culture Area 0.32 cm2
    Array 8 x 12
    Packaging Individually wrapped, 48/Cs.
    Color Black, Clear Bottom
    Material Polystyrene
    No. of Wells 96
    Volume (Metric) Working 75 to 200 μL
    Sterility Sterile
    Surface Treatment Cell Culture
    Well Shape Flat Bottom (F-well)
    For Use With (Application) Attachment and growth of anchorage-dependent cells
    No. per Case 48/Cs.
    No. per Pack 1/Pk.
    Quantity 48/Cs.
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Your feedback has been submitted: Thank you for helping us improve our website.