Learn More
Greiner Bio-One 96-well Chimney designed Round (U) Bottom Microplates
Catalog No. p-7075495
Click to view available options
Color:
Black
Blue
Clear
Green
Natural
Red
White
Yellow
Sterility:
Non-sterile
Sterile
For high-throughput screening in cell based test systems.
  • Non-pyrogenic
  • Conical well design with flat bottom
  • Ensures accurate pipetting
  • Reduces dead volume of well
  • Free of detectable DNase, RNase, human DNA
  • Prevents wicking
TRUSTED_SUSTAINABILITY
Specifications
Breakapart No
Format Standard
Lid Without Lid
Array 8 x 12
Autoclavable Not Autoclavable
Binding Type Non Binding
Color Black
Material Polypropylene
No. of Wells 96
Sterility Non-sterile
Surface Treatment Non-treated
Total Volume (Metric) Well 355 μL
Well Shape Round Bottom (U-well)
Max. RCF 4800 x g (swinging bucket rotor)
No. per Case 100
Quantity 100/Cs.
RNase Free Yes
Well Color Black
Well Design Chimney
Show More Show Less
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.