Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ 96-Well DeepWell™ Polypropylene Microplates
GSA_VAGREENER_CHOICE

Catalog No. 12566121 Shop All Fisherbrand Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Volume (Metric) Well:
1300 μL
2000 μL
Volume (Metric) Working:
1000 μL
1900 μL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility Volume (Metric) Well Volume (Metric) Working
12-566-121 Non-sterile 2000 μL 1900 μL
12-566-611 Sterile 1300 μL 1000 μL
12-566-612 Sterile 2000 μL 1900 μL
12-566-120 Non-sterile 1300 μL 1000 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 12-566-121
Add to Cart
Add to Cart

Ideal for storage because of the low-binding characteristics of polypropylene.

Ideal for storage because of the low-binding characteristics of polypropylene

  • Accommodate even larger working volumes; up to 1.0 mL or 2.0 mL storage volume per well
  • Proteins and DNA will not adhere to PP, allowing for complete sample recovery.
  • Natural color
  • Nonsterile and sterile available

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Array 8 x 12
Color Natural
Material Polypropylene
Volume (Metric) Well 2000 μL
No. of Wells 96
Volume (Metric) Working 1900 μL
Sterility Non-sterile
Well Shape Round
For Use With (Application) For storage of low-binding characteristics of polypropylene
Quantity 60/Cs.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.