Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Biotix™ 96-Well Format Assay Plates
Shop All Biotix Products
Click to view available options
Well Shape:
Conical Bottom (V-well)
Round Bottom (U-well)
Sterility:
Non-sterile
Sterile
Total Volume (Metric) Well:
330 μL
350 μL
Description
- Made from medical grade virgin polypropylene
- DNase, RNase and Endotoxin (pyrogen) free
- Alphanumeric grid to help with sample identification and organization
- Notched bottom corner to be used as a reference for plate orientation
- Available in V-shaped and U-shaped wells

Specifications
Specifications
Breakapart | No |
Format | Standard |
Lid | Without Lid |
Array | 8 x 12 |
Binding Type | Non Binding |
Color | Clear |
Material | Polypropylene |
No. of Wells | 96 |
Sterility | Sterile |
Surface Treatment | Non-treated |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction