Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ 96-Well Low-Profile, Skirted PCR Plates, Natural

Catalog No. p-3599482 Shop All Fisherbrand Products
Click to view available options
Color:
Blue
Green
Natural
Yellow

Catalog No. 14-230-241

Only null left
Add to Cart
Add to Cart

Fit most thermal cyclers

  • Virgin polypropylene
  • Clear well bottom for sample visibility
  • Stackable
  • Raised-rim design
  • Sealable using adhesive films and foils, heat seals, and cap strips
  • 200μL maximum well volume when used with adhesive and heat seals
  • Cut-away corner and alphanumeric grid
  • SBS (Society for Biomolecular Screening) footprint

  • Automation friendly
  • Eight holes in the skirt aid plate positioning and removal from thermal cycler block
  • Cleanroom produced

TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Array 8 x 12
Color Yellow
Product Type PCR Microplate
Skirt Style Skirted
Material Polypropylene
Volume (Metric) Well 200 μL
No. of Wells 96
For Use With (Application) PCR, RT-PCR applications
Rimmed Yes
Well Color Clear
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.