Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BTX™ HT 200 Plate Handler, Auto-Sense Track Switching

Catalog No. btx450401
Click to view available options
:
HT 200; With auto-column switching

Designed to work with 96 well plates. BTX™ Harvard Apparatus 96 Well Plate Handler helps to create a safe and reliable working area for processing electroporation plates.

  • Quickly optimize for better gene delivery
  • Designed to work with 96 well plates
  • Pulse switching technology integrated into package and gold plated contacts to mate with electroporation plate
  • With an adapter to accommodate 25 well plates as well with our ECM 630 generator
  • Prosess up to 96 wells in seconds

Specifications

Format 25-well, 96-well
For Use With (Application) High-Throughput Electroporation
For Use With (Equipment) ECM630, ECM830, Gemini X2
Product Line BTX Electroporation
Product Type Electroporation accessory
Voltage 3000 VDC, 250 VAC
Description HT-200 Microplate Handler, Auto-Sense Track Switching
Display LCD
Electrode Shape Pin
Includes Auto sense track switching, Connectors for standard pulse generator
Pulse Number Range 1 to 99
Pulse Length Range 10 μs to 10 s
Operating Temperature (Metric) 5°C to 40°C
Quantity 1 Ea.
Relative Humidity 20 to 80%
Size (Metric) Gap 2 mm, 4 mm
Weight (Metric) 4.8 kg
Sterility Non-sterile
Connection Type Banana Cables
Interface Digital User
Pulse Form Exponential Decay Wave, Square Wave
High-throughput Compatibility Yes
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.