Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Abgene™ 96-well Sealing Mats for Sample Processing & Storage DeepWell™ and MicroWell Microplates
GSA_VAGREENER_CHOICE

Catalog No. p-3850706
Click to view available options
Temperature Range (Metric):
-20°C to +130°C
-20°C to +55°C
Well Shape:
Round
Square
Fit for Purpose:
PCR Ready

Round or square CapMats, Sealing Mats, MatCaps for sealing 96-well storage plates, available in autoclavable and piercable formats.

Thermo Scientific 96-well Sealing Mats are designed for 96-well storage plates and are available in either piercable or autoclavable formats. The round or square capped mats are made of thermoplastic elastomer and are suitable for long-term storage.

Highlights

  • Certified RNAse, DNAse, PCR Inhibitor, and human DNA free
  • Round or square capped mats for sealing 96-well storage plates
  • Alphanumeric grid-referencing
  • Molded from a proprietary thermoplastic elastomer for a flexible design
  • Suitable for long term storage
  • Available in autoclavable or piercable formats

Pierceable version:

  • Solvent resistant (including DMSO, Ethanol, and Isopropanol)
  • Suitable for Gamma Irradiation
  • Requires 6N for piercing
  • Temperature Range: −20°C to +55°C

Autoclavable version:

  • Suitable for Gamma Irradiation
  • Temperature Range: −20°C to +130°C

 

Hazardous: No
Storage Condition: Ambient

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Color Natural
Solvent Or Alcohol Resistant Alcohol Resistant, Solvent Resistant
Description CapMat for 96-well Polypropylene Sample Processing & Storage Plates
Certifications/Compliance Certified RNase, DNase, PCR Inhibitors, and human DNA Free. ANSI Compliant
Material TPE
Sterility Cleanroom
Autoclavable Autoclavable
No. of Wells 96
No. per Case 50
No. per Pack 10
Plug Diameter 7mm
Temperature Range (English) -4°F to 266°F
For Use With (Application) Designed for sealing 96-well storage plate and suitable for long term storage
Format 96-well
Product Type 96-well capmat, sealing mat
Product Line Abgene
Adhesive None
Certified RNase/DNase-free Yes
Pierceable No
Temperature Range (Metric) -20°C to +130°C
Well Shape Round
Plate Compatibility Abgene 96 Round Well Plate
Fit for Purpose PCR Ready
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.