Learn More
Description
- LoBind material guarantees maximum sample recovery for improved assay results.
- One-piece design: combining a polycarbonate frame and polypropylene wells for optimum performance.
- Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample.
- Exceptionally solid and torque-resistant polycarbonate frame.
- OptiTrack™ matrix for faster sample identification and fewer pipetting errors.
- Raised well rims for effective sealing, also reduces risk of cross-contamination.
- Batch-tested and certified free of DNA, DNAase, RNase and PCR inhibitors (PCR clean).
Specifications
Specifications
| Barcode | Includes Barcode |
| Binding Property | Low DNA Binding |
| Certifications/Compliance | PCR Clean |
| Certified | Yes |
| Format | Standard |
| Lid | With Lid |
| Autoclavable | Autoclavable |
| Packaging | 10 Bags x 30 Plates |
| Product Line | twin.tec |
| Color | Green |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
