Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf™ twin.tec™ PCR Plates LoBind™ - PCR Plates
GREENER_CHOICE

Catalog No. E0030129598 Shop All Eppendorf North America Products
Change view
Click to view available options
Color:
Blue
Clear
Green
Red
Yellow
No. of Wells:
96
384
Skirt Style:
Semi-Skirted
Skirted
Skirted, colorless
Packaging:
10 Bags x 30 Plates
5 Bags x 5 Plates
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color No. of Wells Skirt Style Packaging
E0030129598 Red 96 Skirted 5 Bags x 5 Plates
E0030129512 Clear 96 Skirted 5 Bags x 5 Plates
14-380-052 Clear 96 Skirted 5 Bags x 5 Plates
14-380-057 Clear 96 Skirted, colorless 5 Bags x 5 Plates
E0030129580 Blue 96 Skirted 5 Bags x 5 Plates
E0030129660 Green 96 Skirted 5 Bags x 5 Plates
E0030129679 Yellow 96 Skirted 5 Bags x 5 Plates
E0030129555 Green 96 Skirted 10 Bags x 30 Plates
E0030129563 Yellow 96 Skirted 10 Bags x 30 Plates
14-380-054 Clear 96 Semi-Skirted 5 Bags x 5 Plates
E0030129504 Clear 96 Semi-Skirted 5 Bags x 5 Plates
E0030129547 Clear 384 Skirted 5 Bags x 5 Plates
14-380-053 Clear 384 Skirted 5 Bags x 5 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Catalog No. E0030129598 Supplier Eppendorf™ Supplier No. 0030129598
Add to Cart
Add to Cart

Eppendorf twin.tec™ PCR plates LoBind, available in 96- and 384-well formats, maximize nucleic acid recovery and are certified as PCR clean for consistently reliable results.

  • LoBind material guarantees maximum sample recovery for improved assay results.
  • One-piece design: combining a polycarbonate frame and polypropylene wells for optimum performance.
  • Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample.
  • Exceptionally solid and torque-resistant polycarbonate frame.
  • OptiTrack™ matrix for faster sample identification and fewer pipetting errors.
  • Raised well rims for effective sealing, also reduces risk of cross-contamination.
  • Batch-tested and certified free of DNA, DNAase, RNase and PCR inhibitors (PCR clean).
TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Binding Property Low DNA Binding
Certifications/Compliance PCR Clean
Certified Yes
Format Standard
Lid With Lid
Array 8 x 12
Autoclavable Autoclavable
Packaging 5 Bags x 5 Plates
Product Line twin.tec
Color Red
Disposable Single-use
Product Type PCR Plate
Skirt Style Skirted
Material Polypropylene, Polycarbonate
Volume (Metric) Well 150 μL
No. of Wells 96
Sterility Non-sterile
Well Shape Conical Bottom (V-well)
For Use With (Application) Real Time PCR
High-throughput Compatibility Yes
Max. RCF 6000 x g
No. per Pack 25
Plate Compatibility Yes
Purity or Quality Grade PCR Clean (Non-sterile)
Quantity 25 Pk.
Rimmed Yes
Well Color Colorless
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.