Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Great Lakes Diagnostics™ 96-well V Bottom Plate

Catalog No. 23444013
Click to view available options
Color:
Translucent, Natural

Catalog No. 23-444-013

Supplier: Great Lakes Diagnostics™ 219013

Only null left
Add to Cart
Add to Cart

This 96-well, V bottom polypropylene plate is specifically designed to be compatible with both manual and robotic handling equipment.

This 96-well, V bottom polypropylene plate is specifically designed to be compatible with both manual and robotic handling equipment.

  • Ideal microplate for magnetic particle processing applications
  • Free of DNase and RNase

Specifications

Lid Without Lid
Autoclavable Autoclavable
Product Line KingFisher
Color Translucent, Natural
Disposable Single-use
Product Type Deepwell Plate
For Use With (Equipment) KingFisher™ Extraction and Purification Systems
Skirt Style Non-skirt
Material Polypropylene
Volume (Metric) Well 2.2 mL
No. of Wells 96
Sterility Non-sterile
Well Shape Square, Conical Bottom (V-well)
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.