Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ 968
Click to view available options
Quantity:
10 mg
50 mg
Description
968 is an allosteric inhibitor of glutaminase. Blocks Rho-GTPase-dependent transformation of fibroblasts. Suppresses breast cancer cell growth and invasive activity in vitro. Also inhibits P-493 B lymphoma cell growth and reduces tumor size of P-493 cell xenografts in mice.
Specifications
Specifications
Purity | >99% |
Description | 968 |
Molecular Weight (g/mol) | 475.42 |
Product Type | 968 |
Quantity | 10 mg |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction