Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl 4-aminobenzylphosphonate, 99%

Catalog No. AAB2227906
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2227906 5 g
AAB2227914 25 g
AAB2227922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2227906 Supplier Thermo Scientific Chemicals Supplier No. B2227906
Add to Cart

CAS: 20074-79-7 | C11H18NO3P | 243.24 g/mol

Diethyl 4-aminobenzylphosphonate is used as a reagent in the preparation of [4-(tetradecanoylamino)benzyl]phosphonic acid. It is also used in the production of diethyl p-benzoylaminobenzylphosphonate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl 4-aminobenzylphosphonate is used as a reagent in the preparation of [4-(tetradecanoylamino)benzyl]phosphonic acid. It is also used in the production of diethyl p-benzoylaminobenzylphosphonate.

Solubility
Soluble in methylene chloride, acetone and benzene. Insoluble in water.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20074-79-7
Molecular Formula C11H18NO3P
Molecular Weight (g/mol) 243.24
MDL Number MFCD00007913
InChI Key ZVAYUUUQOCPZCZ-UHFFFAOYSA-N
Synonym diethyl 4-aminobenzylphosphonate, diethyl p-aminobenzylphosphonate, 4-diethoxyphosphorylmethyl aniline, p-aminobenzylphosphonic acid diethyl ester, diethyl p-aminobenzyl phosphonate, phosphonic acid, p-aminobenzyl-, diethyl ester, diethyl 4-aminophenyl methylphosphonate, phosphonic acid, 4-aminophenyl methyl-, diethyl ester, timtec-bb sbb000830
PubChem CID 88358
SMILES CCOP(=O)(CC1=CC=C(N)C=C1)OCC

Specifications

Melting Point 91°C to 94°C
Quantity 5 g
Beilstein 2941613
Solubility Information Soluble in methylene chloride,acetone and benzene. Insoluble in water.
IUPAC Name diethyl [(4-aminophenyl)methyl]phosphonate
Formula Weight 243.24
Percent Purity 99%
Chemical Name or Material Diethyl 4-aminobenzylphosphonate
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 243-504-8
RTECSNumber SZ6480000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.