Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(4-Thiomorpholinyl)aniline, 97%

Catalog No. AAH6336003
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6336003 1 g
AAH63360MD 250 mg
AAH6336006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6336003 Supplier Thermo Scientific Chemicals Supplier No. H6336003
Add to Cart

CAS: 22589-35-1 | C10H14N2S | 194.296 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22589-35-1
Molecular Formula C10H14N2S
Molecular Weight (g/mol) 194.296
MDL Number MFCD01625958
InChI Key RQXQMUUSQGCLPL-UHFFFAOYSA-N
Synonym 4-thiomorpholinoaniline, 4-thiomorpholin-4-yl aniline, 4-thiomorpholin-4-yl phenylamine, 4-thiomorpholinobenzenamine, 4-thiomorpholin-4-yl-phenylamine, benzenamine,4-4-thiomorpholinyl, benzenamine, 4-4-thiomorpholinyl, 4-p-aminophenyl-tetrahydro-4h-1,4-thiazine
PubChem CID 10774206
IUPAC Name 4-thiomorpholin-4-ylaniline
SMILES C1CSCCN1C2=CC=C(C=C2)N

Specifications

Quantity 1 g
UN Number UN2811
Formula Weight 194.3
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-(4-Thiomorpholinyl)aniline
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.