Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Boc-L-tryptophan methyl ester, 96%

Catalog No. AAH6644206
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6644206 5 g
AAH6644214 25 g
AAH6644222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6644206 Supplier Thermo Scientific Chemicals Supplier No. H6644206
Add to Cart

CAS: 33900-28-6 | C17H22N2O4 | 318.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33900-28-6
Molecular Formula C17H22N2O4
Molecular Weight (g/mol) 318.37
MDL Number MFCD01075094
InChI Key QXLOVPXUZAOKBL-AWEZNQCLSA-N
Synonym boc-trp-ome, boc-l-trp-ome, boc-l-tryptophan methyl ester, s-methyl 2-tert-butoxycarbonyl amino-3-1h-indol-3-yl propanoate, l-tryptophan,n-1,1-dimethylethoxy carbonyl-, methyl ester, n-boc-tryptophan methyl ester, methyl 2s-2-tert-butoxycarbonyl amino-3-1h-indol-3-yl propanoate, ambotzbaa5960, n-boc-l-tryptophan methyl ester
PubChem CID 7021503
IUPAC Name methyl (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-(1H-indol-3-yl)propanoate
SMILES COC(=O)[C@H](CC1=CNC2=CC=CC=C12)NC(=O)OC(C)(C)C

Specifications

Color White
Quantity 5 g
Formula Weight 318.37
Percent Purity 96%
Physical Form Powder
Chemical Name or Material N-Boc-L-tryptophan methyl ester
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.