Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,9-Dibromononane, 97%, Thermo Scientific Chemicals

Catalog No. AC112870250
Change view
Click to view available options
Quantity:
5 g
25 g
50 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC112870250 25 g
AC112870050 5 g
AC112870500 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC112870250 Supplier Thermo Scientific Chemicals Supplier No. 112870250
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4549-33-1
InChI Key WGAXVZXBFBHLMC-UHFFFAOYSA-N
Synonym nonane, 1,9-dibromo, nonamethylene dibromide, nonamethylene bromide, 1,9-dibromnonane, nonane,9-dibromo, 1,9-dibromononane, pubchem3900, acmc-1ai7y, ksc236g2b
PubChem CID 20677
IUPAC Name 1,9-dibromononane
SMILES C(CCCCBr)CCCCBr

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.4000g/mL
Boiling Point 285°C to 288°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C9H18Br2
Refractive Index 1.4950 to 1.497
Linear Formula Br(CH2)9Br
Quantity 25 g
Beilstein 01, 166
Specific Gravity 1.4
Molecular Weight (g/mol) 286.04
Formula Weight 286.04
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1, 9-Dibromononane, 97%
Show More Show Less
EINECSNumber 224-913-0
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.