Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tributyl borate, 98%

Catalog No. AC177195000
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC177195000 500 mL Glass bottle
AC177191000 100 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC177195000 Supplier Thermo Scientific Chemicals Supplier No. 177195000
Add to Cart

CAS: 688-74-4 | C12H27BO3 | 230.155 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 688-74-4
InChI Key LGQXXHMEBUOXRP-UHFFFAOYSA-N
Synonym n-butyl borate, tri-n-butyl borate, tributoxyborane, butyl borate, tributoxyboron, boron tributoxide, borester 2, tri-n-butoxyborane, tris butoxy borane, borane, tributoxy
PubChem CID 12712
IUPAC Name tributyl borate
SMILES B(OCCCC)(OCCCC)OCCCC

Specifications

Melting Point -70.0°C
Color Colorless
Density 0.8530g/mL
Boiling Point 230.0°C to 235.0°C
Flash Point 93°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C12H27BO3
Refractive Index 1.4080 to 1.4100
Linear Formula (CH3CH2CH2CH2O)3B
Quantity 500 mL
Beilstein 01,II,398
Fieser 13,326
Specific Gravity 0.853
Solubility Information Solubility in water: decomposes. Other solubilities: miscible with most common organic liquids
Molecular Weight (g/mol) 230.15
Formula Weight 230.15
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Tributyl borate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
EINECSNumber 211-706-5

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.