Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Furaldehyde, 99%

Catalog No. AC181100025
Change view
Click to view available options
Quantity:
25 mL
250 mL
1 L
2.5 L
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC181100025 2.5 L Glass bottle
AC181100250 25 mL Glass bottle
AC181102500 250 mL Glass bottle
AC181100010 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC181100025 Supplier Thermo Scientific Chemicals Supplier No. 181100025
Add to Cart

CAS: 98-01-1 | C5H4O2 | 96.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-01-1
Molecular Formula C5H4O2
Molecular Weight (g/mol) 96.09
MDL Number MFCD00003229
InChI Key HYBBIBNJHNGZAN-UHFFFAOYSA-N
Synonym furfural, 2-furaldehyde, 2-furancarboxaldehyde, furaldehyde, 2-formylfuran, furfuraldehyde, fural, 2-furanaldehyde, 2-furancarbonal, 2-furfural
PubChem CID 7362
ChEBI CHEBI:34768
IUPAC Name furan-2-carbaldehyde
SMILES O=CC1=CC=CO1

Specifications

Acidity 0.02 eq/L max.
CAS Min % 98.5
CAS Max % 100.0
Melting Point -37°C
Color Amber to Brown
Density 1.1600g/mL
Boiling Point 159°C to 161°C
Flash Point 60°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5250 to 1.527
Quantity 2.5 L
Residue after Evaporation 0.5% max.
Specific Gravity 1.16
Merck Index 15,4333
Solubility Information Solubility in water: 83g/L (20°C). Other solubilities: soluble in alcohol,ether and benzene
Viscosity 1.49 mPa.s (25°C)
Formula Weight 96.08
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Furaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable liquid and vapour.
Toxic if swallowed.
Harmful in contact with skin.
Fatal if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Sus
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to f
EINECSNumber 202-627-7
Recommended Storage This product darkens on contact with air

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.