Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(triphenylphosphine)palladium(II) diacetate, 99%

Catalog No. AC209270050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC209270050 5 g Glass bottle
AC209270010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC209270050 Supplier Thermo Scientific Chemicals Supplier No. 209270050
Add to Cart

CAS: 14588-08-0 | C40H36O4P2Pd | 749.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14588-08-0
Molecular Formula C40H36O4P2Pd
Molecular Weight (g/mol) 749.09
MDL Number MFCD00010013
InChI Key YOUIUHDWQIUKAO-UHFFFAOYSA-L
Synonym bis triphenylphosphinepalladium acetate
PubChem CID 73357379
IUPAC Name palladium(2+);triphenylphosphanium;diacetate
SMILES CC(=O)O[Pd++]OC(C)=O.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 136.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 14% min. (Pd)
Packaging Glass bottle
Linear Formula [(C6H5)3P]2Pd(O2CCH3)2
Quantity 5 g
Formula Weight 749.08
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Bis(triphenylphosphine)palladium(II) diacetate, 99%
Show More Show Less
EINECSNumber 238-628-4

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.