Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Heptadecanol, 97%

Catalog No. AC221230050
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC221230050 5 g
AC221230250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC221230050 Supplier Thermo Scientific Chemicals Supplier No. 221230050
Add to Cart

CAS: 1454-85-9 | C17H36O | 256.474 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1454-85-9
InChI Key GOQYKNQRPGWPLP-UHFFFAOYSA-N
Synonym 1-heptadecanol, heptadecyl alcohol, heptadecanol, n-heptadecanol, unii-n3il85tmcx, 1-hydroxyheptadecane, alcohols, c16-18, n3il85tmcx, heptadecanol mixed primary isomers, n-heptadecanol-1
PubChem CID 15076
ChEBI CHEBI:77470
IUPAC Name heptadecan-1-ol
SMILES CCCCCCCCCCCCCCCCCO

Specifications

Melting Point 51.0°C to 56.0°C
Color White
Boiling Point 308.5°C
Flash Point 154°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C17H36O
Linear Formula CH3(CH2)16OH
Quantity 5 g
Beilstein 01,I,22
Solubility Information Solubility in water: slightly soluble in water.
Molecular Weight (g/mol) 256.46
Formula Weight 256.46
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 1-Heptadecanol
Show More Show Less
EINECSNumber 215-932-5

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.