Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Dibromotetrafluorobenzene, 99%

Catalog No. AC311350050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC311350050 5 g Glass bottle
AC311350010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC311350050 Supplier Thermo Scientific Chemicals Supplier No. 311350050
Add to Cart

CAS: 827-08-7 | C6Br2F4 | 307.88 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 827-08-7
Molecular Formula C6Br2F4
Molecular Weight (g/mol) 307.88
MDL Number MFCD00039209
InChI Key IPLUWQPTPKNBRD-UHFFFAOYSA-N
Synonym 1,2-dibromotetrafluorobenzene, 3,4,5,6-tetrafluoro-1,2-dibromobenzene, tetrafluoro-1,2-dibromobenzene, benzene, 1,2-dibromotetrafluoro, benzene, 1,2-dibromo-3,4,5,6-tetrafluoro, 1,2-dibromoperfluorobenzene, dtfb, pubchem2195, dibromotetrafluorobenzene, acmc-20ap6i
PubChem CID 70007
IUPAC Name 1,2-dibromo-3,4,5,6-tetrafluorobenzene
SMILES C1(=C(C(=C(C(=C1F)Br)Br)F)F)F

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Density 2.2300g/mL
Boiling Point 198.0°C
Flash Point 98°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5150 to 1.5170
Linear Formula Br2C6F4
Quantity 5 g
Specific Gravity 2.23
Formula Weight 307.88
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1, 2-Dibromotetrafluorobenzene
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 212-564-7

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.