Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(3R,4R)-(-)-1-Benzyl-3,4-pyrrolidindiol, 97%

Catalog No. AC348720050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC348720050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC348720050 Supplier Thermo Scientific Chemicals Supplier No. 348720050
Add to Cart

CAS: 163439-82-5 | C11H15NO2 | 193.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 163439-82-5
MDL Number MFCD01073894
InChI Key QJRIUWQPJVPYSO-GHMZBOCLSA-N
Synonym 3r,4r-1-benzylpyrrolidine-3,4-diol, 3r,4r---1-benzyl-3,4-pyrrolidindiol, 3r,4r---1-benzyl-3,4-pyrrolidinediol, 3,4-pyrrolidinediol, 1-phenylmethyl-, 3r,4r, pubchem14982, trans-3,4-dihydroxy-n-benzylpyrrolidine, 3r,4r-1-benzyl-3,4-pyrrolidinediol, 3r-1-benzylpyrrolidine-3beta,4alpha-diol, 3r,4r-rel-1-phenylmethyl-3,4-pyrrolidinediol, 3,4-pyrrolidinediol,1-phenylmethyl-, 3r,4r
PubChem CID 7021491
IUPAC Name (3R,4R)-1-benzylpyrrolidine-3,4-diol
SMILES C1C(C(CN1CC2=CC=CC=C2)O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 95.0°C to 101.0°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C11H15NO2
Quantity 5 g
Specific Rotation Condition −32° (20°C c=1,MeOH)
Specific Rotation − 32.00
Molecular Weight (g/mol) 193.24
Formula Weight 193.24
Percent Purity 97%
Physical Form Powder, Crystals or Chunks
Chemical Name or Material (3R, 4R)-(-)-1-Benzyl-3, 4-pyrrolidindiol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.