Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-1-propanol, 97%

Catalog No. AC349700050
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC349700050 5 mL Glass bottle
AC349700250 25 mL Glass bottle
AC349701000 100 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC349700050 Supplier Thermo Scientific Chemicals Supplier No. 349700050
Add to Cart

CAS: 627-18-9 | C3H7BrO | 138.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 627-18-9
Molecular Formula C3H7BrO
Molecular Weight (g/mol) 138.99
MDL Number MFCD00002942
InChI Key RQFUZUMFPRMVDX-UHFFFAOYSA-N
Synonym 3-bromo-1-propanol, trimethylene bromohydrin, 1-propanol, 3-bromo, 3-bromopropanol, 3-hydroxypropyl bromide, 1-bromo-3-propanol, 3-bromo-propan-1-ol, ccris 5979, 1-bromo-3-hydroxypropane, unii-3t4axh68fh
PubChem CID 12308
IUPAC Name 3-bromopropan-1-ol
SMILES OCCCBr

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.6000g/mL
Boiling Point 62.0°C (5.0 mmHg)
Flash Point 65°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4860 to 1.4900
Linear Formula Br(CH2)3OH
Quantity 5 mL
Beilstein 01,356
Specific Gravity 1.6
Solubility Information Solubility in water: 167g/L water (20°C). Other solubilities: slightly soluble in ethanol and ether, soluble in most solvents
Formula Weight 138.99
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Bromo-1-propanol, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 210-986-6
RTECSNumber UA7385000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.