Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-6-chloro-4-picoline, 98%

Catalog No. AC360990010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC360990010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC360990010 Supplier Thermo Scientific Chemicals Supplier No. 360990010
Add to Cart

CAS: 66909-38-4 | C6H7ClN2 | 142.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66909-38-4
Molecular Formula C6H7ClN2
Molecular Weight (g/mol) 142.59
InChI Key PLSMBLIGZJWLEJ-UHFFFAOYSA-N
Synonym 3-amino-6-chloro-4-picoline, 5-amino-2-chloro-4-methylpyridine, 5-amino-2-chloro-4-picoline, 3-amino-6-chloro-4-methyl-pyridine, 3-pyridinamine, 6-chloro-4-methyl, pubchem6206, 6-chloro-4-methyl-3-pyridinamine, 6-chloro-4-methyl-3-pyridylamine, 3-amino-6-chloro-4-methylpyridine, 6-chloro-4-methyl-3-pyridineamine
PubChem CID 2734425
IUPAC Name 6-chloro-4-methylpyridin-3-amine
SMILES CC1=CC(=NC=C1N)Cl

Specifications

Melting Point 69.0°C to 73.0°C
Color Bordeaux to Brown
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 142.59
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 3-Amino-6-chloro-4-picoline
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.