Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indoine Blue, Thermo Scientific™

Catalog No. AC365330050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC365330050 5 g
AC365330010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC365330050 Supplier Thermo Scientific Chemicals Supplier No. 365330050
Add to Cart

Chemical Identifiers

CAS 4569-88-4
Molecular Formula C30H24ClN5O
Molecular Weight (g/mol) 506.01
MDL Number MFCD00011924
InChI Key SCMDRBZEIUMBBQ-UHFFFAOYSA-N
PubChem CID 9575841
SMILES [Cl-].CC1=CC2=NC3=CC(C)=C(N)C=C3[N+](C3=CC=CC=C3)=C2C=C1N\N=C1/C(=O)C=CC2=CC=CC=C12

Specifications

Infrared Spectrum Authentic
IUPAC Name 3-amino-2,8-dimethyl-7-{2-[(1Z)-2-oxo-1,2-dihydronaphthalen-1-ylidene]hydrazin-1-yl}-5-phenyl-5λ⁵-phenazin-5-ylium chloride
Lambda Max 595 to 601nm (0.001% in methanol)
Formula Weight 506.01
Loss on Drying 10% max. (110°C, 1 h)
Packaging Glass bottle
Quantity 5 g
Chemical Name or Material Indoine Blue

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.