Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hematein

Catalog No. AC370840050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC370840050 5 g Glass bottle
AC370840250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC370840050 Supplier Thermo Scientific Chemicals Supplier No. 370840050
Add to Cart

CAS: 475-25-2 | C16H12O6 | 300.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 475-25-2
Molecular Formula C16H12O6
Molecular Weight (g/mol) 300.27
MDL Number MFCD00036187
InChI Key HLUCICHZHWJHLL-UHFFFAOYNA-N
Synonym hematein, haematein, hematine, hydroxybrasilein, benz b indeno 1,2-d pyran-9 6h-one, 6a,7-dihydro-3,4,6a,10-tetrahydroxy, 3,4,6a,10-tetrahydroxy-6,7-dihydroindeno 2,1-c chromen-9-one, 3,4,6a,10-tetrahydroxy-6a,7-dihydrobenz b indeno 1,2-d pyran-9 6h-one, 6a,7-dihydro-3,4,6a,10-tetrahydroxybenz b indeno 1,2-d pyran-9 6h-one, hematein,haematein,hydroxybrazilein,hydroxybrasilein,c.i.75290, hydroxybrazilein
PubChem CID 10138
IUPAC Name 3,4,6a,10-tetrahydroxy-6,7-dihydroindeno[2,1-c]chromen-9-one
SMILES OC1=CC=C2C3=C4C=C(O)C(=O)C=C4CC3(O)COC2=C1O

Specifications

Melting Point 180°C
Color Brown
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 5 g
Beilstein 18,IV,3343
Merck Index 15,4669
Solubility Information Solubility in water: very slightly soluble. Other solubilities: slightly solubel in ethanol,ether,soluble in alkaline solutions,insoluble in benzene,chloroform
Formula Weight 300.26
Physical Form Crystalline Powder
Chemical Name or Material Hematein
Show More Show Less
EINECSNumber 207-492-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.