Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-bromo-n-methylaniline, 98%

Catalog No. AC387360050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC387360050 5 g Glass bottle
AC387360010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC387360050 Supplier Thermo Scientific Chemicals Supplier No. 387360050
Add to Cart

CAS: 66584-32-5 | C7H8BrN | 186.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66584-32-5
Molecular Formula C7H8BrN
Molecular Weight (g/mol) 186.05
MDL Number MFCD05664376
InChI Key HKOSFZXROYRVJT-UHFFFAOYSA-N
Synonym n-methyl-3-bromoaniline, 3-bromo-n-methyl aniline, benzenamine, 3-bromo-n-methyl, acmc-20an89, 3-bromophenyl-methylamine, 3-bromo-phenyl-methyl-amine, 3-bromo-n-methylaniline
PubChem CID 7018299
IUPAC Name 3-bromo-N-methylaniline
SMILES CNC1=CC(=CC=C1)Br

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow to Brown
Density 1.4610g/mL
Boiling Point 253°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.6110 to 1.6130 (20°C, 589nm)
Linear Formula BrC6H4NHCH3
Quantity 5 g
Specific Gravity 1.461
Formula Weight 186.05
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Bromo-N-methylaniline
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.