Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Bis(dimethylamino)benzil, 98%

Catalog No. AC402390010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC402390010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC402390010 Supplier Thermo Scientific Chemicals Supplier No. 402390010
Add to Cart

CAS: 17078-27-2 | C18H20N2O2 | 296.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17078-27-2
Molecular Formula C18H20N2O2
Molecular Weight (g/mol) 296.36
InChI Key AVFUVYIDYFXFSX-UHFFFAOYSA-N
Synonym 4,4'-bis dimethylamino benzil, 1,2-bis 4-dimethylamino phenyl ethane-1,2-dione, ethandione, bis 4-dimethylamino phenyl, bis 4-dimethylamino phenyl ethane-1,2-dione, acmc-209e1j, benzil-based compound, 35, bis n,n-dimethylamino benzil, 4,4/'-bis dimethylamino benzil, bis 4-dimethylamino phenyl diketone
PubChem CID 222570
IUPAC Name 1,2-bis[4-(dimethylamino)phenyl]ethane-1,2-dione
SMILES CN(C)C1=CC=C(C=C1)C(=O)C(=O)C2=CC=C(C=C2)N(C)C

Specifications

Melting Point 200.0°C to 204.0°C
Color Green to Green-Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 296.36
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4, 4'-Bis(dimethylamino)benzil

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.