Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methylsulphonylbenzoic acid, 95%, Thermo Scientific™

Catalog No. AC424860025
Change view
Click to view available options
Quantity:
2.5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC424860025 2.5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC424860025 Supplier Thermo Scientific Chemicals Supplier No. 424860025
Add to Cart

Chemical Identifiers

CAS 5345-27-7
Molecular Formula C8H8O4S
Molecular Weight (g/mol) 200.22
MDL Number MFCD00216488
InChI Key KUTBMATZUQWFSR-UHFFFAOYSA-N
Synonym 3-methylsulfonyl benzoic acid, 3-methylsulphonyl benzoic acid, 3-methylsulphonylbenzoic acid, 3-methanesulfonylbenzoic acid, 3-methanesulfonyl-benzoic acid, benzoic acid, 3-methylsulfonyl, 3-mesylbenzoic acid, pubchem10870, acmc-1auq1
PubChem CID 220380
IUPAC Name 3-methylsulfonylbenzoic acid
SMILES CS(=O)(=O)C1=CC=CC(=C1)C(=O)O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 230°C to 238°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Linear Formula SO2CH3C6H4COOH
Quantity 2.5 g
Formula Weight 200.22
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 3-Methylsulphonylbenzoic acid, 95%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.