Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Aminopyridine-3-carboxaldehyde, 95%

Catalog No. AC437350010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC437350010 1 g
AC437350050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC437350010 Supplier Thermo Scientific Chemicals Supplier No. 437350010
Add to Cart

CAS: 42373-30-8 | C6H7N2O | 123.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42373-30-8
Molecular Formula C6H7N2O
Molecular Weight (g/mol) 123.13
MDL Number MFCD04038972
InChI Key GTPZHMGXKZIHKW-UHFFFAOYSA-O
Synonym 4-aminonicotinaldehyde, 4-amino-3-formylpyridine, 4-amino-pyridine-3-carbaldehyde, 4-aminopyridine-3-carboxaldehyde, 4-amino-3-pyridinecarboxaldehyde, 4-amino-pyridine-3-carboxaldehyde, 3-pyridinecarboxaldehyde, 4-amino, zlchem 837, pubchem7681, acmc-1aqvh
PubChem CID 2761033
IUPAC Name 4-aminopyridine-3-carbaldehyde
SMILES NC1=CC=[NH+]C=C1C=O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 111.2°C to 112.7°C
Color Cream to White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 122.13
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 4-Aminopyridine-3-carboxaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.