Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

H-2K[k] Mouse anti-Mouse, FITC, Clone: 36-7-5, BD

Catalog No. BDB553592 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB553592 0.5 mg
BDB562005 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB553592 Supplier BD Biosciences Supplier No. 553592
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 36-7-5 antibody reacts with the H-2K[k] MHC class I alloantigen. Cross-reactivity with splenocytes of SJL/Hsd mice has not been observed by flow cytometric analysis. Reactivity with other haplotypes (e.g., b, d, q) has not been observed. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Mouse
Clone: 36-7-5
Isotype: IgG2a κ
Species Reactivity: Mouse
Immunogen: A.AL mouse splenocytes

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen H-2K[k]
Applications Flow Cytometry
Classification Monoclonal
Clone 36-7-5
Concentration 0.5mg/mL
Conjugate FITC
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen A.AL mouse splenocytes
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.