Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD178 Hamster anti-Mouse, NA/LE, Unlabeled, Clone: MFL3, BD

Catalog No. BDB555290 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555290 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555290 Supplier BD Biosciences Supplier No. 555290
Add to Cart
Add to Cart

Hamster Monoclonal Antibody

The MFL3 antibody reacts with CD178 (Fas Ligand, CD95 Ligand) on all strains tested. In the mouse, Fas Ligand is expressed on activated T cell lines and in spleen, testis, and eye. FasL mRNA has been demonstrated at various levels in bone marrow, thymus, spleen, lymph node, lung, small intestine, testis, and uterus. Moreover, T-cell activators, but not B-cell activators, enhanced the expression of FasL mRNA in splenocytes; and FasL mRNA was restricted to the T-cell lineage among a panel of cell lines from lymphoid tissues. Fas Ligand is not functional in mice homozygous for the gld (generalized lympho-proliferative disease) mutation; these mice cannot limit the expansion of activated lymphocytes and develop autoimmune disease. Fas Ligand is a member of the TNF/NGF family, which binds to CD95 (Fas), inducing apoptotic cell death. This Fas/Fas Ligand interaction is believed to participate in T-cell development, the regulation of immune responses, and cell-mediated cytotoxic mechanisms. There is mounting evidence that Fas Ligand is also proinflammatory, mediating neutrophil extravasation and chemotaxis. Fas Ligand is released from the surface of transfectant cells by metalloproteinases, and the soluble Fas Ligand may block the activities of the membrane-bound molecule. The MFL3 mAb has been reported to efficiently inhibit the cytotoxicity of mouse Fas Ligand-transfected cells against human Fas-transfected cells. This hamster mAb to a mouse leukocyte antigen does not cross-react with rat leukocytes.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD178
Applications Flow Cytometry
Classification Monoclonal
Clone MFL3
Concentration 1.0mg/mL
Conjugate Unconjugated
Description Fas Ligand, CD95 Ligand
Formulation No azide/low endotoxin: Aqueous buffered solution containing no preservative, 0.2μm sterile filtered. Endotoxin level is ≤0.01 EU/μg (≤0.001 ng/μg) of protein as determined by the LAL assay.
Host Species Hamster
Immunogen Mouse FasL-transfected cells
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.