Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD18 Mouse anti-Human, FITC, Clone: 6.7, BD

Catalog No. BDB555923 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
50 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555923 100 Tests
BDB557156 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB555923 Supplier BD Biosciences Supplier No. 555923
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 6.7 monoclonal antibody specifically binds to β2 integrin, a 90-95 kDa transmembrane protein, which associates non-covalently as a heterodimer with the a chains of CD11a, CD11b, or CD11c. CD18 is expressed on lymphocytes, monocytes, and more weakly on granulocytes. LFA-1 has been shown to play an important role in homotypic and heterotypic cellular adhesion in immune and inflammatory responses.

This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD18
Applications Flow Cytometry
Classification Monoclonal
Clone 6.7
Conjugate FITC
Description ITGB2; Integrin beta-2; Beta subunit for LFA-1, MAC-1/CR3, and p150,95
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Cloned Human DS6 TCR γδ+ Cells
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Cynomolgus, Human, Pig, Rabbit, Rhesus Monkey
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.