Learn More
Description
Reacts with a 50 - 58 kDa type I glycoprotein molecule of the killer cell inhibitory receptor (KIR) family of proteins. CD158b has also been known as GLI83, p50.2, or p58.2. CD158b is composed of two extracellular Ig-like domains. It is expressed on a subset of NK cells and it may also be present on rare T cells (either CD3/TCR αβ+, or CD3/TCR γδ+). CD158b regulates NK cell mediated cytolytic activity by interacting with HLA-C alleles (Cw 1, 3, 7 and 8).
Specifications
Specifications
| Antigen | CD158b |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | CH-L |
| Conjugate | FITC |
| Formulation | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Host Species | Mouse |
| Purification Method | Affinity Purified |
| Quantity | 100 Tests |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.