Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher BioReagents™ Lysozyme, Egg White (White Cryst. Powder)

Catalog No. BP5351 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP535-1 1 g
BP535-5 5 g
BP535-25 25 g
BP535-10 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. BP535-1
Add to Cart
This item is not returnable. View return policy
  • Application: This enzyme is used to lyse E. coli for preparation of plasmid DNA from transformed cell cultures
  • Not on TSCA inventory: for research and development use only; not for manufacturing or commercial purposes
  • Enzyme Activity: ≈20,000U/mg dry weight
  • Solubility: Readily soluble at 5mg/mL in 0.1M KH2PO4 to give colorless solution
TRUSTED_SUSTAINABILITY

Specifications

Color White
For Use With (Application) Used to lyse E. coli for preparation of plasmid DNA from transformed cell cultures
Content And Storage <0°C
Source Egg white
Quantity 1 g
Product Type Lysozyme
Form Crystalline Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.